Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3S,4R)-4-(4'-Fluorophenyl)-3-hydroxymethyl-1-methylpiperidine, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
105812-81-5
C13H18FNO
223.291
MFCD06658161
CXRHUYYZISIIMT-AAEUAGOBSA-N
3s,4r-4-4-fluorophenyl-3-hydroxymethyl-1-methylpiperidine,3s,4r-4-4-fluorophenyl-1-methyl-3-piperidinemethanol,paroxol,unii-idr7471br7,3s,4r-4-4-fluorophenyl-1-methylpiperidin-3-yl methanol,3s-4-4'-fluorophenyl-3-hydroxymethyl-1-methylpiperidine,3s,4r-4-4-fluoro-phenyl-1-methyl-piperidin-3-yl-methanol,3-piperidinemethanol, 4-4-fluorophenyl-1-methyl-, 3s,4r,trans-4-4-fluorophenyl-3-hydroxymethyl-1-methyl-piperidine
2734218
[(3S,4R)-4-(4-fluorophenyl)-1-methylpiperidin-3-yl]methanol
CN1CCC(C(C1)CO)C2=CC=C(C=C2)F
105812-81-5
C13H18FNO
223.291
MFCD06658161
CXRHUYYZISIIMT-AAEUAGOBSA-N
3s,4r-4-4-fluorophenyl-3-hydroxymethyl-1-methylpiperidine,3s,4r-4-4-fluorophenyl-1-methyl-3-piperidinemethanol,paroxol,unii-idr7471br7,3s,4r-4-4-fluorophenyl-1-methylpiperidin-3-yl methanol,3s-4-4'-fluorophenyl-3-hydroxymethyl-1-methylpiperidine,3s,4r-4-4-fluoro-phenyl-1-methyl-piperidin-3-yl-methanol,3-piperidinemethanol, 4-4-fluorophenyl-1-methyl-, 3s,4r,trans-4-4-fluorophenyl-3-hydroxymethyl-1-methyl-piperidine
2734218
[(3S,4R)-4-(4-fluorophenyl)-1-methylpiperidin-3-yl]methanol
CN1CCC(C(C1)CO)C2=CC=C(C=C2)F
Color White
Infrared Spectrum Authentic
Specific Rotation Condition − 38.00 (20.00°C c=1,MeOH)
Packaging Glass bottle
Melting Point 97°C to 101°C
Quantity 5g
Specific Rotation -35° to -41° (20°C, 589nm) (c=1, MeOH)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 223.29
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material (3S, 4R)-4-(4'-Fluorophenyl)-3-hydroxymethyl-1-methylpiperidine
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

TSCA : TSCA