Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Polypropylene Isolation Gown
Offers protection from splashes and spills. Fisherbrand™ Polypropylene Isolation Gown features elastic wrist cuffs and easy neck and waist ties.
This item is not returnable.
View return policy
Description
- Durable; Spunbound polypropylene, Elastic wrist cuff, Easy neck and waist ties, Provides protection from common dirt, dust and grime, minor splashes and particulates
- Spunbond polypropylene
- Elastic wrist cuff
- Easy neck and waist ties
- Provides protection from common dirt, dust and grime, minor splashes and particulates
Specifications
Yellow | |
53 in. | |
Polypropylene | |
Non-sterile | |
10.24 in. |
Industry and Lab/clean room/Medical Area | |
34 cm | |
3X-Large | |
Isolation gown | |
26 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title