Alfa Aesar™ 4-(1,4-Cyclohexadien-1-yl)-1-butanol, 97%, stab. with 0.1% BHT

Manufacturer: Alfa Aesar™ H3605806

Quantity 5g

 View more versions of this product

Catalog No. AAH3605806

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C10H16O
    Formula Weight 152.23
    MDL Number MFCD22682825
    Synonym 4-Cyclohexa-1,4-dienyl-1-butanol
    Chemical Name or Material 4-(1,4-Cyclohexadien-1-yl)-1-butanol, stabilized with 0.1% BHT

    Specifications

    Formula Weight 152.23
    Molecular Formula C10H16O
    MDL Number MFCD22682825
    Synonym 4-Cyclohexa-1,4-dienyl-1-butanol
    Recommended Storage 2° to 8°C
    Flash Point 103°C (217°F)
    Boiling Point 247° to 248°C
    Quantity 5g
    Chemical Name or Material 4-(1,4-Cyclohexadien-1-yl)-1-butanol, stabilized with 0.1% BHT
    Assay 97%

    2° to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit