Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(1-Cyclohexen-1-yl)morpholine, 97%

Click to view available options
Quantity:
25g
100g
670-80-4
C10H17NO
167.252
MFCD00006163
IIQFBBQJYPGOHJ-UHFFFAOYSA-N
4-cyclohex-1-enyl-morpholine,1-morpholino-1-cyclohexene,1-morpholinocyclohexene,n-morpholino-1-cyclohexene,4-1-cyclohexen-1-yl morpholine,morpholine, 4-1-cyclohexen-1-yl,4-cyclohex-1-en-1-yl morpholine,4-1-cyclohexenyl morpholine,1-morpholin-1-ylcyclohexene,1-n-morpholino cyclohexene
69589
4-(cyclohexen-1-yl)morpholine
C1CCC(=CC1)N2CCOCC2
670-80-4
C10H17NO
167.252
MFCD00006163
IIQFBBQJYPGOHJ-UHFFFAOYSA-N
4-cyclohex-1-enyl-morpholine,1-morpholino-1-cyclohexene,1-morpholinocyclohexene,n-morpholino-1-cyclohexene,4-1-cyclohexen-1-yl morpholine,morpholine, 4-1-cyclohexen-1-yl,4-cyclohex-1-en-1-yl morpholine,4-1-cyclohexenyl morpholine,1-morpholin-1-ylcyclohexene,1-n-morpholino cyclohexene
69589
4-(cyclohexen-1-yl)morpholine
C1CCC(=CC1)N2CCOCC2
Boiling Point 118°C to 120°C (10mmHg)
Quantity 100g
Flash Point 68°C (154°F)
Beilstein 118696
Formula Weight 167.25
Percent Purity 97%
Chemical Name or Material 4-(1-Cyclohexen-1-yl)morpholine

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Recommended Storage : 2° to 8°C