Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(1-Oxo-5Z,8Z,11Z,14Z-eicosatetraenylamino)butanoic acid, 98%

Click to view available options
Quantity:
5mg
10mg
128201-89-8
C24H39NO3
389.58
MFCD05863981
JKUDIEXTAYKJNX-CGRWFSSPSA-N
N-Arachidonyl-GABA; NAGABA
44634744
4-[[(5E,8E,11E,14E)-icosa-5,8,11,14-tetraenoyl]amino]butanoic acid
CCCCCC=CCC=CCC=CCC=CCCCC(=O)NCCCC(=O)O
128201-89-8
C24H39NO3
389.58
MFCD05863981
JKUDIEXTAYKJNX-CGRWFSSPSA-N
N-Arachidonyl-GABA; NAGABA
44634744
4-[[(5E,8E,11E,14E)-icosa-5,8,11,14-tetraenoyl]amino]butanoic acid
CCCCCC=CCC=CCC=CCC=CCCCC(=O)NCCCC(=O)O
Quantity 5mg
Color Yellow
Sensitivity Air Sensitive
Formula Weight 389.58
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 4-(1-Oxo-5Z,8Z,11Z,14Z-eicosatetraenylamino)butanoic acid

storageNote1 : -30° to -10°C