Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4'-(1-Piperazinyl)acetophenone, 94%

Click to view available options
Quantity:
5g
25g
100g
51639-48-6
C12H16N2O
204.273
MFCD00005962
KPXVKKBJROCIJB-UHFFFAOYSA-N
1-4-acetylphenyl piperazine,1-4-piperazin-1-yl phenyl ethanone,4'-piperazinoacetophenone,1-4-piperazin-1-ylphenyl ethanone,4-piperazinoacetophenone,piperazin-4-ylacetophenone,1-4-piperazin-1-yl-phenyl-ethanone,p-piperazinoacetophenone,1-4-piperazin-1-yl-phenyl ethanone
97269
1-(4-piperazin-1-ylphenyl)ethanone
CC(=O)C1=CC=C(C=C1)N2CCNCC2
51639-48-6
C12H16N2O
204.273
MFCD00005962
KPXVKKBJROCIJB-UHFFFAOYSA-N
1-4-acetylphenyl piperazine,1-4-piperazin-1-yl phenyl ethanone,4'-piperazinoacetophenone,1-4-piperazin-1-ylphenyl ethanone,4-piperazinoacetophenone,piperazin-4-ylacetophenone,1-4-piperazin-1-yl-phenyl-ethanone,p-piperazinoacetophenone,1-4-piperazin-1-yl-phenyl ethanone
97269
1-(4-piperazin-1-ylphenyl)ethanone
CC(=O)C1=CC=C(C=C1)N2CCNCC2
Quantity 5g
Melting Point 93°C to 98°C
UN Number UN3259
Formula Weight 204.27
Percent Purity 94%
Chemical Name or Material 4'-(1-Piperazinyl)acetophenone

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.