Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 4'-(1-Piperazinyl)acetophenone, 94%
Click to view available options
Quantity:
5g
25g
100g
Specifications
Melting Point | 93°C to 98°C |
Quantity | 100g |
UN Number | UN3259 |
Formula Weight | 204.27 |
Percent Purity | 94% |
Chemical Name or Material | 4'-(1-Piperazinyl)acetophenone |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
EINECSNumber : 257-332-6
TSCA : No
Recommended Storage : Ambient temperatures