Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(1-Piperazinyl)indole, 95%

Click to view available options
Quantity:
250mg
1g
84807-09-0
C12H15N3
201.273
MFCD00831776
YZKSXUIOKWQABW-UHFFFAOYSA-N
4-1-piperazinyl-1h-indole,4-piperazin-1-yl-1h-indole,4-piperazino-1h-indole,1h-indole, 4-1-piperazinyl,4-piperazino indole,1-4-indolyl piperazine,1h-indole-4-1-piperazinyl dihydrochloride,4-piperazinoindole,4-piperazino-indole,pubchem22668
594590
4-piperazin-1-yl-1H-indole
C1CN(CCN1)C2=CC=CC3=C2C=CN3
84807-09-0
C12H15N3
201.273
MFCD00831776
YZKSXUIOKWQABW-UHFFFAOYSA-N
4-1-piperazinyl-1h-indole,4-piperazin-1-yl-1h-indole,4-piperazino-1h-indole,1h-indole, 4-1-piperazinyl,4-piperazino indole,1-4-indolyl piperazine,1h-indole-4-1-piperazinyl dihydrochloride,4-piperazinoindole,4-piperazino-indole,pubchem22668
594590
4-piperazin-1-yl-1H-indole
C1CN(CCN1)C2=CC=CC3=C2C=CN3
Quantity 250mg
Beilstein 9198621
Formula Weight 201.27
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 4-(1-Piperazinyl)indole

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : NM1302400