Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(1-Pyrrolidinyl)piperidine, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
5004-07-9
C9H18N2
154.25
MFCD00038011
STWODXDTKGTVCJ-UHFFFAOYSA-N
4-1-pyrrolidinyl piperidine,4-pyrrolidin-1-yl piperidine,4-pyrrolidin-1-yl-piperidine,4-pyrrolidinopiperidine,4-1-pyrrolidinyl-piperidine,piperidine, 4-1-pyrrolidinyl,4-pyrrolidinylpiperidine,4-pyrrolizinopiperidine,4-pyrrolidino-piperidine
78703
4-pyrrolidin-1-ylpiperidine
C1CCN(C1)C2CCNCC2
5004-07-9
C9H18N2
154.25
MFCD00038011
STWODXDTKGTVCJ-UHFFFAOYSA-N
4-1-pyrrolidinyl piperidine,4-pyrrolidin-1-yl piperidine,4-pyrrolidin-1-yl-piperidine,4-pyrrolidinopiperidine,4-1-pyrrolidinyl-piperidine,piperidine, 4-1-pyrrolidinyl,4-pyrrolidinylpiperidine,4-pyrrolizinopiperidine,4-pyrrolidino-piperidine
78703
4-pyrrolidin-1-ylpiperidine
C1CCN(C1)C2CCNCC2
Quantity 25g
Melting Point 52.0°C to 57.0°C
Color White
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 154.25
Physical Form Crystals
Percent Purity 99%
Chemical Name or Material 4-(1-Pyrrolidinyl)piperidine
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.