Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(1-Pyrrolidinyl)pyridine, 98%

Click to view available options
Quantity:
5g
25g
100g
2456-81-7
C9H12N2
148.209
MFCD00006415
RGUKYNXWOWSRET-UHFFFAOYSA-N
4-pyrrolidinopyridine,4-pyrrolidin-1-yl pyridine,4-1-pyrrolidinyl pyridine,pyridine, 4-1-pyrrolidinyl,4-pyrrolidin-1-yl-pyridine,4-pyrrolidinylpyridine,1-4-pyridyl pyrrolidine,zlchem 148,4-pyrrolidinepyridine,4-pyrrollidinopyridine
75567
4-pyrrolidin-1-ylpyridine
C1CCN(C1)C2=CC=NC=C2
2456-81-7
C9H12N2
148.209
MFCD00006415
RGUKYNXWOWSRET-UHFFFAOYSA-N
4-pyrrolidinopyridine,4-pyrrolidin-1-yl pyridine,4-1-pyrrolidinyl pyridine,pyridine, 4-1-pyrrolidinyl,4-pyrrolidin-1-yl-pyridine,4-pyrrolidinylpyridine,1-4-pyridyl pyrrolidine,zlchem 148,4-pyrrolidinepyridine,4-pyrrollidinopyridine
75567
4-pyrrolidin-1-ylpyridine
C1CCN(C1)C2=CC=NC=C2
Boiling Point 171°C to 173°C (21mmHg)
Quantity 100g
Flash Point 142°C (287°F)
Melting Point 54°C to 58°C
UN Number UN2923
Beilstein 472443
Formula Weight 148.21
Percent Purity 98%
Chemical Name or Material 4-(1-Pyrrolidinyl)pyridine

GHS H Statement
H300-H314-H318
Fatal if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.