Learn More
4-1BB Ligand/TNFSF9 Rabbit anti-Human, Alexa Fluor™ 750, Clone: 2357A, R&D Systems™
Rabbit Monoclonal Antibody
Manufacturer:  R&D Systems FAB22952S100UG
Catalog No. FB22952S100
Add to cart
This item is not returnable. View return policy
Description
Description
4-1BB Ligand/TNFSF9 Monoclonal antibody specifically detects 4-1BB Ligand/TNFSF9 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
4-1BB Ligand/TNFSF9
2357A
Flow Cytometry 0.25-1 ug/10^6 cells
P41273.1
4-1BB ligand, 41BB Ligand, 4-1BBL, 4-1BB-L, CD137L, homolog of mouse 4-1BB-L, receptor 4-1BB ligand, TNFSF9, tumor necrosis factor (ligand) superfamily, member 9, tumor necrosis factor ligand superfamily member 9
E. coli-derived human 4-1BB Ligand/TNFSF9, Arg71-Glu254, Accession No. P41273.1
Protein A or G purified from cell culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human 4-1BB Ligand/TNFSF9 in direct ELISA.
Flow Cytometry
Alexa Fluor™ 750
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide.
Purified, Supernatant, Conjugated
Rabbit
IgG
100 μg
Primary
8744.0
Human