Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-1BB Ligand/TNFSF9, Rabbit anti-Mouse, Clone: TKS-1, Novus Biologicals™

Rabbit Monoclonal Antibody

$156.50 - $419.00

Specifications

Antigen 4-1BB Ligand/TNFSF9
Clone TKS-1
Applications Flow Cytometry, Blocking Assay
Host Species Rabbit
Target Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB016424
View Documents
Novus Biologicals
NBP2811150.025MG
0.025 mg
Each for $156.50
Add to cart
 
NB016425
View Documents
Novus Biologicals
NBP2811150.2MG
0.2 mg
Each for $419.00
Add to cart
 
Description

Description

4-1BB Ligand/TNFSF9 Monoclonal antibody specifically detects 4-1BB Ligand/TNFSF9 in Mouse Samples. It is validated for Flow Cytometry, Blocking Assay.
Specifications

Specifications

4-1BB Ligand/TNFSF9
Flow Cytometry, Blocking Assay
Mouse
Primary
TKS-1
Rabbit
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit