Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-1BB/TNFRSF9/CD137 Rabbit anti-Mouse, Clone: LOB12.3, Novus Biologicals
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP275201
Description
4-1BB/TNFRSF9/CD137 Monoclonal antibody specifically detects 4-1BB/TNFRSF9/CD137 in Mouse samples. It is validated for Flow Cytometry.Specifications
4-1BB/TNFRSF9/CD137 | |
LOB12.3 | |
Flow Cytometry 1:10 - 1:1000, Functional | |
Purified | |
TNFRSF9 | |
This antibody was generated by immunizing rats with recombinant CD137-Fc fusion protein. | |
Protein A purified | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
Monoclonal | |
This antibody binds to mouse 4-1BB (CD137). |
Flow Cytometry, Functional Assay | |
Unconjugated | |
0.02% Proclin 300 | |
4-1BB, 4-1BB ligand receptor, CD137 antigen, CD137MGC2172, CDw137, FLJ43501, ILAhomolog of mouse 4-1BB, induced by lymphocyte activation (ILA), interleukin-activated receptor, homolog of mouse Ly63, receptor protein 4-1BB, T cell antigen ILA, T-cell antigen 4-1BB homolog, T-cell antigen ILA, tumor necrosis factor receptor superfamily member 9, tumor necrosis factor receptor superfamily, member 9 | |
Rabbit | |
IgG Kappa | |
0.2 mg | |
Primary | |
3604.0 | |
Mouse |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title