Learn More
Novus Biologicals™ 4-1BB/TNFRSF9/CD137 ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP191278
Catalog No. NBP191278
Add to cart
Description
Description
The Human 4-1BB/TNFRSF9/CD137 ELISA Kit (Colorimetric) measures human 4-1BB/TNFRSF9/CD137 in cell culture supernatant, serum.
Specifications
Specifications
0.78 - 50ng/mL
3604
ELISA Kit
0.2ng/mL
Storage of components varies. See protocol for specific instructions.
Apoptosis, Immune System Diseases, Immunology
4-1BB, 4-1BB ligand receptor, CD137 antigen, CD137MGC2172, CDw137, FLJ43501, ILAhomolog of mouse 4-1BB, induced by lymphocyte activation (ILA), interleukin-activated receptor, homolog of mouse Ly63, receptor protein 4-1BB, T cell antigen ILA, T-cell antig
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sCD137, Biotin-Conjugate anti-human sCD137 polyclonal antibody, Streptavidin-HRP, Human sCD137 Standard lyophilized, 100ng/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum
4-1BB/TNFRSF9/CD137
TNFRSF9