Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-1BB/TNFRSF9/CD137 Goat anti-Rat, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

$119.60 - $379.00

Specifications

Antigen 4-1BB/TNFRSF9/CD137
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Format Affinity Purified
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AF9029SP
View Documents Promotion Details
R&D Systems
AF9029SP
25 ug
Each for $119.60
Add to cart
 
AF9029100
View Documents Promotion Details
R&D Systems
AF9029100
100 ug
Each for $379.00
Add to cart
 
Description

Description

4-1BB/TNFRSF9/CD137 Polyclonal specifically detects 4-1BB/TNFRSF9/CD137 in Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications

Specifications

4-1BB/TNFRSF9/CD137
Affinity Purified
Unconjugated
IgG
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Flow Cytometry, Immunocytochemistry, CyTOF
Goat
Antigen Affinity-purified
RUO
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit