Learn More
4-1BB/TNFRSF9/CD137 Mouse anti-Human, Alexa Fluor 700, Clone: MM0071-9G43, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP211954AF700
Catalog No. NB211954700
Description
Description
4-1BB/TNFRSF9/CD137 Monoclonal antibody specifically detects 4-1BB/TNFRSF9/CD137 in Human samples. It is validated for Western Blot.
Specifications
Specifications
4-1BB/TNFRSF9/CD137
MM0071-9G43
Western Blot
Purified
TNFRSF9
Human CD137
Protein G purified
Apoptosis, Immune System Diseases, Immunology
Primary
3604
Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
4-1BB, 4-1BB ligand receptor, CD137 antigen, CD137MGC2172, CDw137, FLJ43501, ILAhomolog of mouse 4-1BB, induced by lymphocyte activation (ILA), interleukin-activated receptor, homolog of mouse Ly63, receptor protein 4-1BB, T cell antigen ILA, T-cell antigen 4-1BB homolog, T-cell antigen ILA, tumor necrosis factor receptor superfamily member 9, tumor necrosis factor receptor superfamily, member 9
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human