Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-1BB/TNFRSF9/CD137 Mouse anti-Human, Baboon, Primate, Janelia Fluor™ 549, Clone: 4B4-1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NB10077887JF549

Catalog No. N107787J549

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

4-1BB/TNFRSF9/CD137 Monoclonal antibody specifically detects 4-1BB/TNFRSF9/CD137 in Human, Baboon, Primate samples. It is validated for Flow Cytometry, ELISA.
Specifications

Specifications

4-1BB/TNFRSF9/CD137
4B4-1
Flow Cytometry, ELISA
Conjugated, Purified
TNFRSF9
Ectodomain of recombinant human 4-1BB fusion protein
Protein A or G purified
Apoptosis, Immune System Diseases, Immunology
Primary
3604.0
Baboon, Human, Primate
ELISA, Flow Cytometry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
4-1BB, 4-1BB ligand receptor, CD137 antigen, CD137MGC2172, CDw137, FLJ43501, ILAhomolog of mouse 4-1BB, induced by lymphocyte activation (ILA), interleukin-activated receptor, homolog of mouse Ly63, receptor protein 4-1BB, T cell antigen ILA, T-cell antigen 4-1BB homolog, T-cell antigen ILA, tumor necrosis factor receptor superfamily member 9, tumor necrosis factor receptor superfamily, member 9
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
CD137 (4B4-1)
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit