Learn More
4-1BB/TNFRSF9/CD137, Rabbit anti-Human, Alexa Fluor 594, Clone: 2356C, R&D Systems™
Rabbit Monoclonal Antibody
Manufacturer:  R&D Systems FAB8382T100UG
Catalog No. FAB8382T100
Add to cart
This item is not returnable. View return policy
Description
Description
4-1BB/TNFRSF9/CD137 Monoclonal antibody specifically detects 4-1BB/TNFRSF9/CD137 in Human Samples. It is validated for Flow Cytometry.
Specifications
Specifications
4-1BB/TNFRSF9/CD137
2356C
Flow Cytometry 0.25-1 ug/10^6 cells
Q07011
41BB, 4-1BB, 4-1BB ligand receptor, CD137, CD137 antigen, CD137MGC2172, CDw137, FLJ43501, ILA, ILAhomolog of mouse 4-1BB, induced by lymphocyte activation (ILA), interleukin-activated receptor, homolog of mouse Ly63, receptor protein 4-1BB, T cell antigen ILA, T-cell antigen 4-1BB homolog, T-cell antigen ILA, TNFRSF9, tumor necrosis factor receptor superfamily member 9, tumor necrosis factor receptor superfamily, member 9
Chinese Hamster Ovary cell line CHO-derived human 4-1BB/TNFRSF9/CD137, Leu24-His183, Accession # Q07011
Protein A or G purified from cell culture supernatant
RUO
Primary
3604
Human
Flow Cytometry
Alexa Fluor 594
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified
Rabbit
IgG
100 ug
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human 4-1BB/TNFRSF9/CD137 in direct ELISAs.