Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Imidazol-1-yl)phenol, 97%, Acros Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
10041-02-8
C9H8N2O
160.176
MFCD00005281
CYKCUAPYWQDIKR-UHFFFAOYSA-N
4-imidazol-1-yl phenol,4-1h-imidazol-1-yl phenol,4-imidazol-1-yl-phenol,4-1-imidazolyl phenol,1-p-hydroxyphenyl imidazole,phenol, 4-1h-imidazol-1-yl,1-4-hydroxyphenyl imidazole,4-imidazolylphenol,4‐ 1h‐imidazol‐1‐yl phenol compound 1,p-1-imidazolyl phenol
82315
4-imidazol-1-ylphenol
C1=CC(=CC=C1N2C=CN=C2)O
10041-02-8
C9H8N2O
160.176
MFCD00005281
CYKCUAPYWQDIKR-UHFFFAOYSA-N
4-imidazol-1-yl phenol,4-1h-imidazol-1-yl phenol,4-imidazol-1-yl-phenol,4-1-imidazolyl phenol,1-p-hydroxyphenyl imidazole,phenol, 4-1h-imidazol-1-yl,1-4-hydroxyphenyl imidazole,4-imidazolylphenol,4‐ 1h‐imidazol‐1‐yl phenol compound 1,p-1-imidazolyl phenol
82315
4-imidazol-1-ylphenol
C1=CC(=CC=C1N2C=CN=C2)O
Melting Point 204.0°C to 207.0°C
Infrared Spectrum Authentic
Quantity 5g
Color Beige-Brown to Brown-Gray
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 160.18
Physical Form Fine Crystalline Powder
Percent Purity 97%
Chemical Name or Material 4-(Imidazol-1-yl)phenol
Show More Show Less

GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning