Alfa Aesar™ 4-(2,2,2-Trimethylacetamido)pyridine, 97%

Manufacturer: Alfa Aesar™ H3156409

Quantity 10g

 View more versions of this product

Catalog No. AAH3156409

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 70298-89-4
    Molecular Formula C10H14N2O
    Formula Weight 178.24
    MDL Number MFCD00996248
    Synonym 2,2-Dimethyl-N-(4-pyridyl)propionamide; 4-(Pivaloylamino)pyridine
    Chemical Name or Material 4-(2,2,2-Trimethylacetamido)pyridine

    Specifications

    Formula Weight 178.24
    Molecular Formula C10H14N2O
    MDL Number MFCD00996248
    CAS 70298-89-4
    Synonym 2,2-Dimethyl-N-(4-pyridyl)propionamide; 4-(Pivaloylamino)pyridine
    Melting Point 135° to 137°C
    Quantity 10g
    Chemical Name or Material 4-(2,2,2-Trimethylacetamido)pyridine
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit