Alfa Aesar™ 4-[(2,4-Dimethoxyphenyl)(Fmoc-amino)methyl]phenoxyacetic acid, 98+%

Manufacturer: Alfa Aesar™ H5487406

Quantity 5g

 View more versions of this product

Catalog No. AAH5487406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 145069-56-3
    Molecular Formula C32H29NO7
    Formula Weight 539.58
    MDL Number MFCD00153509
    Synonym Rink amide linker
    Chemical Name or Material 4-[(2,4-Dimethoxyphenyl)(Fmoc-amino)methyl]phenoxyacetic acid

    Specifications

    Formula Weight 539.58
    Molecular Formula C32H29NO7
    Recommended Storage 2° to 8°C
    Quantity 5g
    MDL Number MFCD00153509
    CAS 145069-56-3
    Synonym Rink amide linker
    Melting Point ≈180°C
    Chemical Name or Material 4-[(2,4-Dimethoxyphenyl)(Fmoc-amino)methyl]phenoxyacetic acid
    Assay ≥98%

    2° to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit