Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Aminoethyl)-1-benzylpiperidine 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
86945-25-7
C14H22N2
218.344
MFCD02093423
OUYRPOHWEJUTCQ-UHFFFAOYSA-N
2-(1-Benzyl-4-piperidyl)ethylamine
1867002
2-(1-benzylpiperidin-4-yl)ethanamine
C1CN(CCC1CCN)CC2=CC=CC=C2
86945-25-7
C14H22N2
218.344
MFCD02093423
OUYRPOHWEJUTCQ-UHFFFAOYSA-N
2-(1-Benzyl-4-piperidyl)ethylamine
1867002
2-(1-benzylpiperidin-4-yl)ethanamine
C1CN(CCC1CCN)CC2=CC=CC=C2
Color Yellow
Quantity 25g
Formula Weight 218.34
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 4-(2-Aminoethyl)-1-benzylpiperidine