Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AEBSF Hydrochloride, 4-(2-Aminoethyl)-benzenesulfonyl Fluoride HCl, White Powder, Fisher BioReagents
Click to view available options
Quantity:
10 mg
50 mg
100 mg
500 mg
1 g
Packaging:
Amber Glass
Description
- Irreverisble serine protease inhibitor
- Inhibits proteases like trypsin, chymotrypsin, kallikrein, plasmin, and thrombin
- Acts as Sulfonyl fluoride that reacts with hydroxy group of the active site serine residue
- More soluble and less toxic
Specifications
Melting Point | 175°C |
Color | White |
Infrared Spectrum | Pass Test |
Assay Percent Range | ≥97 % |
Packaging | Amber Glass |
Quantity | 500 mg |
Formula Weight | 239.7 |
Percent Purity | 97% min. |
Physical Form | Powder or Solid |
Chemical Name or Material | 4-(2-Aminoethyl)benzenesulfonyl Fluoride Hydrochloride |
Safety and Handling
Hazard Category | Signal Word: DANGER! |
Precautionary Statement | Emergency Overview Corrosive. Irritating to eyes and skin. May cause burns. NFPA Health: 3 Flammability: |
TSCA | Not on TSCA inventory: for R and D use only; not for manufacturing or commercial purposes. |
Recommended Storage | RT, protect from light. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction