Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Aminoethyl)morpholine, TCI America™

Click to view available options
Quantity:
25mL
100mL
500mL
2038-03-1
C6H14N2O
130.19
MFCD00006182
RWIVICVCHVMHMU-UHFFFAOYSA-N
4-2-aminoethyl morpholine,2-morpholinoethanamine,4-morpholineethanamine,2-morpholinoethylamine,n-aminoethylmorpholine,2-morpholin-4-yl ethan-1-amine,morpholine, 4-2-aminoethyl,n-2-aminoethyl morpholine,n-2-aminoethylmorpholine,2-morpholin-4-ylethylamine
408285
2-morpholin-4-ylethanamine
C1COCCN1CCN
2038-03-1
C6H14N2O
130.19
MFCD00006182
RWIVICVCHVMHMU-UHFFFAOYSA-N
4-2-aminoethyl morpholine,2-morpholinoethanamine,4-morpholineethanamine,2-morpholinoethylamine,n-aminoethylmorpholine,2-morpholin-4-yl ethan-1-amine,morpholine, 4-2-aminoethyl,n-2-aminoethyl morpholine,n-2-aminoethylmorpholine,2-morpholin-4-ylethylamine
408285
2-morpholin-4-ylethanamine
C1COCCN1CCN
Quantity 25mL
Color White-Yellow
Boiling Point 205°C
UN Number 2923
Formula Weight 130.19
Physical Form Crystal-Lump at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 4-(2-Aminoethyl)morpholine

RTECSNumber : QD7350000

TSCA : Yes