Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Aminoethyl)tetrahydropyran, 97%, Acros Organics™

Click to view available options
Quantity:
250mg
1g
65412-03-5
C7H15NO
129.2
BZMADPOGYCRPAI-UHFFFAOYSA-N
4-2-aminoethyl tetrahydropyran,4-2-aminoethyl tetrahydro-2h-pyran,2-oxan-4-yl ethan-1-amine,2-oxan-4-yl ethanamine,2-tetrahydro-2h-pyran-4-yl ethanamine,4-aminoethyl-tetrahydropyrane,2h-pyran-4-ethanamine, tetrahydro,4-aminoethyl tetrahydropyran,2-tetrahydro-pyran-4-yl-ethylamine,2-tetrahydro-pyran-4-yl ethylamine
2773198
2-(oxan-4-yl)ethanamine
C1COCCC1CCN
65412-03-5
C7H15NO
129.2
BZMADPOGYCRPAI-UHFFFAOYSA-N
4-2-aminoethyl tetrahydropyran,4-2-aminoethyl tetrahydro-2h-pyran,2-oxan-4-yl ethan-1-amine,2-oxan-4-yl ethanamine,2-tetrahydro-2h-pyran-4-yl ethanamine,4-aminoethyl-tetrahydropyrane,2h-pyran-4-ethanamine, tetrahydro,4-aminoethyl tetrahydropyran,2-tetrahydro-pyran-4-yl-ethylamine,2-tetrahydro-pyran-4-yl ethylamine
2773198
2-(oxan-4-yl)ethanamine
C1COCCC1CCN
Packaging Glass Bottle
Boiling Point 88.0°C to 89.0°C (13.0mmHg)
Infrared Spectrum Authentic
Quantity 1g
Flash Point 74°C
Assay Percent Range 96% min. (GC)
Formula Weight 129.2
Percent Purity 97%
Chemical Name or Material 4-(2-Aminoethyl)tetrahydropyran

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning