Learn More
Alfa Aesar™ 4-(2-Chlorophenoxy)benzenesulfonyl chloride, 96%
Manufacturer:  Alfa Aesar™ H32346MD
Quantity 250mg
 View more versions of this product
Catalog No. AAH32346MD
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
610277-84-4
303.17
UN3261
C12H8Cl2O3S
MFCD01631899
4-(2-Chlorophenoxy)benzenesulfonyl chloride
Specifications
250mg
C12H8Cl2O3S
MFCD01631899
4-(2-Chlorophenoxy)benzenesulfonyl chloride
610277-84-4
303.17
UN3261
96%
Safety and Handling
SDS & Safety Documents
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.