Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(2-Cyano-3-ethoxy-3-oxo-1-propen-1-yl)benzeneboronic acid pinacol ester

Manufacturer:  Alfa Aesar™ H5113006

Quantity 5g

 View more versions of this product

Catalog No. AAH5113006


 

Due to product restrictions, please to purchase or view availability for this product.


Specifications Description & Specifications

Chemical Identifiers

C18H22BNO4
MFCD11113044
4-(2-Cyano-3-ethoxy-3-oxo-1-propen-1-yl)benzeneboronic acid pinacol ester
327.18
Ethyl alpha-cyano-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cinnamate

Specifications

5g
327.18
Ethyl alpha-cyano-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cinnamate
C18H22BNO4
MFCD11113044
4-(2-Cyano-3-ethoxy-3-oxo-1-propen-1-yl)benzeneboronic acid pinacol ester

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P280f
Wear protective clothing.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit