Alfa Aesar™ 4-(2-Cyano-3-ethoxy-3-oxo-1-propen-1-yl)benzeneboronic acid pinacol ester

Manufacturer: Alfa Aesar™ H5113014

Quantity 25g

 View more versions of this product

Catalog No. AAH5113014

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    Molecular Formula C18H22BNO4
    Formula Weight 327.18
    MDL Number MFCD11113044
    Synonym Ethyl alpha-cyano-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cinnamate
    Chemical Name or Material 4-(2-Cyano-3-ethoxy-3-oxo-1-propen-1-yl)benzeneboronic acid pinacol ester

    Specifications

    Formula Weight 327.18
    Molecular Formula C18H22BNO4
    MDL Number MFCD11113044
    Synonym Ethyl alpha-cyano-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cinnamate
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    Health Hazard 3 GHS P Statement
    P280f
    Wear protective clothing.
    Quantity 25g
    Chemical Name or Material 4-(2-Cyano-3-ethoxy-3-oxo-1-propen-1-yl)benzeneboronic acid pinacol ester

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P280f
    Wear protective clothing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit