Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Furyl)-3-buten-2-one, 95%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
623-15-4
C8H8O2
136.15
MFCD00039566
GBKGJMYPQZODMI-SNAWJCMRSA-N
furfurylideneacetone,4-2-furyl-3-buten-2-one,3-buten-2-one, 4-2-furanyl,fam polymer,oramin r,furfural acetone,oramin special gr,furfuralacetone polymer,polyfurfurylidineacetone,furfuralacetone
735940
(E)-4-(furan-2-yl)but-3-en-2-one
CC(=O)C=CC1=CC=CO1
623-15-4
C8H8O2
136.15
MFCD00039566
GBKGJMYPQZODMI-SNAWJCMRSA-N
furfurylideneacetone,4-2-furyl-3-buten-2-one,3-buten-2-one, 4-2-furanyl,fam polymer,oramin r,furfural acetone,oramin special gr,furfuralacetone polymer,polyfurfurylidineacetone,furfuralacetone
735940
(E)-4-(furan-2-yl)but-3-en-2-one
CC(=O)C=CC1=CC=CO1
Quantity 5g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Orange to Brown
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 136.15
Physical Form Liquid After Melting
Percent Purity ≥94%
Chemical Name or Material 4-(2-Furyl)-3-buten-2-one, 95%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

LIGHT SENSITIVE

EINECSNumber : 210-774-3

RTECSNumber : EM9939000

TSCA : TSCA