Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-[2-Hydroxyethyl(methyl)amino]benzeneboronic acid pinacol ester, 95%

Manufacturer:  Alfa Aesar™ H5194103

Quantity 1g

 View more versions of this product

Catalog No. AAH5194103


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1206641-45-3
291.2
2-(Methyl[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]amino)ethanol
C16H26BNO3
MFCD16294544
4-[2-Hydroxyethyl(methyl)amino]benzeneboronic acid pinacol ester

Specifications

1g
C16H26BNO3
MFCD16294544
4-[2-Hydroxyethyl(methyl)amino]benzeneboronic acid pinacol ester
1206641-45-3
291.2
2-(Methyl[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]amino)ethanol
95%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit