Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Keto-1-benzimidazolinyl)piperidine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
20662-53-7
C12H15N3O
217.27
BYNBAMHAURJNTR-UHFFFAOYSA-N
4-2-keto-1-benzimidazolinyl piperidine,1-piperidin-4-yl-1h-benzo d imidazol-2 3h-one,4-2-keto-1-benzimidozolinyl piperidine,unii-4rp0aed2pc,1-piperidin-4-yl-1h-benzimidazol-2-ol,4rp0aed2pc,1-4-piperidinyl-1,3-dihydro-2h-benzimidazol-2-one,1-piperidin-4-yl-2,3-dihydro-1h-1,3-benzodiazol-2-one,1-piperidin-4-yl-3h-1,3-benzodiazol-2-one,1,3-dihydro-1-4-piperidinyl-2h-benzimidazol-2-one
88638
3-piperidin-4-yl-1H-benzimidazol-2-one
C1CNCCC1N2C3=CC=CC=C3NC2=O
20662-53-7
C12H15N3O
217.27
BYNBAMHAURJNTR-UHFFFAOYSA-N
4-2-keto-1-benzimidazolinyl piperidine,1-piperidin-4-yl-1h-benzo d imidazol-2 3h-one,4-2-keto-1-benzimidozolinyl piperidine,unii-4rp0aed2pc,1-piperidin-4-yl-1h-benzimidazol-2-ol,4rp0aed2pc,1-4-piperidinyl-1,3-dihydro-2h-benzimidazol-2-one,1-piperidin-4-yl-2,3-dihydro-1h-1,3-benzodiazol-2-one,1-piperidin-4-yl-3h-1,3-benzodiazol-2-one,1,3-dihydro-1-4-piperidinyl-2h-benzimidazol-2-one
88638
3-piperidin-4-yl-1H-benzimidazol-2-one
C1CNCCC1N2C3=CC=CC=C3NC2=O
Color White to Yellow
Melting Point 183.0°C to 185.0°C
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
Assay Percent Range 97%
Solubility Information Solubility in water: 3g/L (20°C). Other solubilities: 32g/100mL methanol- 13g/100mL ethanol,7g/100mL 2-propanol- 0.75g/100mL acetone,3.1g/100mL methylene chloride- 9.4g/100mL n,n-dmf
Formula Weight 217.27
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 4-(2-Keto-1-benzimidazolinyl)piperidine
Show More Show Less

GHS Signal Word: Warning

EINECSNumber : 243-950-3