Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(2-Methoxyethyl)phenol, 98%

Click to view available options
Quantity:
10g
50g
250g
56718-71-9
C9H12O2
152.193
MFCD00017537
FAYGEALAEQKPDI-UHFFFAOYSA-N
4-2-methoxyethyl phenol,p-2-methoxyethyl phenol,phenol, 4-2-methoxyethyl,4-methoxyethylphenol,unii-py1w0akw8i,metoprolol impurity b,metoprolol impurity i,py1w0akw8i,4-hydroxyphenylethyl methyl ether
92516
4-(2-methoxyethyl)phenol
COCCC1=CC=C(C=C1)O
56718-71-9
C9H12O2
152.193
MFCD00017537
FAYGEALAEQKPDI-UHFFFAOYSA-N
4-2-methoxyethyl phenol,p-2-methoxyethyl phenol,phenol, 4-2-methoxyethyl,4-methoxyethylphenol,unii-py1w0akw8i,metoprolol impurity b,metoprolol impurity i,py1w0akw8i,4-hydroxyphenylethyl methyl ether
92516
4-(2-methoxyethyl)phenol
COCCC1=CC=C(C=C1)O
Melting Point 43°C to 45°C
Quantity 250g
Formula Weight 152.19
Percent Purity 98%
Chemical Name or Material 4-(2-Methoxyethyl)phenol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning