Alfa Aesar™ 4-(2-Methyl-3-furylthio)-5-nonanone, 95%

Manufacturer: Alfa Aesar™ H27560MC

Quantity 100mg

 View more versions of this product

Catalog No. AAH27560MC

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 61295-50-9
    Molecular Formula C14H22O2S
    Formula Weight 254.39
    MDL Number MFCD08741453
    Chemical Name or Material 4-(2-Methyl-3-furylthio)-5-nonanone

    Specifications

    Formula Weight 254.39
    MDL Number MFCD08741453
    Quantity 100mg
    Molecular Formula C14H22O2S
    CAS 61295-50-9
    Chemical Name or Material 4-(2-Methyl-3-furylthio)-5-nonanone
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit