Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(2-Naphthyl)phenol, 98%

Click to view available options
Quantity:
250mg
1g
5g
6336-82-9
C16H12O
220.271
MFCD18312886
NIRHUNSXEDESLN-UHFFFAOYSA-N
4-naphthalen-2-yl phenol,4-naphthalen-2-yl-phenol,4-2-naphthyl phenol,d0l3ax,2-4-hydroxyphenyl naphthalene
236868
4-naphthalen-2-ylphenol
C1=CC=C2C=C(C=CC2=C1)C3=CC=C(C=C3)O
6336-82-9
C16H12O
220.271
MFCD18312886
NIRHUNSXEDESLN-UHFFFAOYSA-N
4-naphthalen-2-yl phenol,4-naphthalen-2-yl-phenol,4-2-naphthyl phenol,d0l3ax,2-4-hydroxyphenyl naphthalene
236868
4-naphthalen-2-ylphenol
C1=CC=C2C=C(C=CC2=C1)C3=CC=C(C=C3)O
Quantity 250mg
Beilstein 2524079
Formula Weight 220.27
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 4-(2-Naphthyl)phenol