Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(2-Thienyl)-3-buten-2-one, 97%

Click to view available options
Quantity:
1g
5g
25g
874-83-9
C8H8OS
152.211
MFCD00053051
CIMALVIHZVKKPE-SNAWJCMRSA-N
4-2-thienyl but-3-en-2-one,trans-4-2-thienyl-3-buten-2-one,4-2-thienyl-3-buten-2-one,3e-4-thiophen-2-yl but-3-en-2-one,4-thiophen-2-yl but-3-en-2-one,e-4-thiophen-2-ylbut-3-en-2-one,4-2-thienyl-3-buten-2-one mixt. of cis and trans,3e-4-2-thienyl but-3-en-2-one,3-buten-2-one,4-2-thienyl,3-buten-2-one,4-2-thienyl-, 3e
735949
(E)-4-thiophen-2-ylbut-3-en-2-one
CC(=O)C=CC1=CC=CS1
874-83-9
C8H8OS
152.211
MFCD00053051
CIMALVIHZVKKPE-SNAWJCMRSA-N
4-2-thienyl but-3-en-2-one,trans-4-2-thienyl-3-buten-2-one,4-2-thienyl-3-buten-2-one,3e-4-thiophen-2-yl but-3-en-2-one,4-thiophen-2-yl but-3-en-2-one,e-4-thiophen-2-ylbut-3-en-2-one,4-2-thienyl-3-buten-2-one mixt. of cis and trans,3e-4-2-thienyl but-3-en-2-one,3-buten-2-one,4-2-thienyl,3-buten-2-one,4-2-thienyl-, 3e
735949
(E)-4-thiophen-2-ylbut-3-en-2-one
CC(=O)C=CC1=CC=CS1
Melting Point 22°C to 25°C
Boiling Point 86°C to 88°C (1mmHg)
Quantity 25g
Formula Weight 152.22
Percent Purity 97%
Chemical Name or Material 4-(2-Thienyl)-3-buten-2-one