Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-(2-Thienyl)benzaldehyde, 97%, Thermo Scientific™
Click to view available options
1g; Amber Glass Bottle
Specifications
CAS | 107834-03-7 |
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Molecular Formula | C11H8OS |
MDL Number | MFCD02682003 |
Quantity | 1g |
Synonym | 4-2-thienyl benzaldehyde, 4-thiophen-2-yl benzaldehyde, 4-thiophen-2-yl-benzaldehyde, benzaldehyde,4-2-thienyl, 4-2-thienyl benzaldeyde, acmc-209vfl, 2-4-formylphenyl thiophene, 4-thien-2-yl benzaldehyde, benzaldehyde, 4-2-thienyl |
InChI Key | UECDQUOWFRTJOH-UHFFFAOYSA-N |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction