Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Thienyl)benzenesulfonyl chloride, 95%, Acros Organics

Click to view available options
Quantity:
1g
5g
209592-91-6
C10H7ClO2S2
258.75
IRDZCMUQNGEQEP-UHFFFAOYSA-N
4-2-thienyl benzenesulfonyl chloride,benzenesulfonyl chloride, 4-2-thienyl,4-thiophen-2-yl benzene-1-sulfonyl chloride,4-thiophen-2-yl benzenesulfonyl chloride
18387759
4-thiophen-2-ylbenzenesulfonyl chloride
C1=CSC(=C1)C2=CC=C(C=C2)S(=O)(=O)Cl
209592-91-6
C10H7ClO2S2
258.75
IRDZCMUQNGEQEP-UHFFFAOYSA-N
4-2-thienyl benzenesulfonyl chloride,benzenesulfonyl chloride, 4-2-thienyl,4-thiophen-2-yl benzene-1-sulfonyl chloride,4-thiophen-2-yl benzenesulfonyl chloride
18387759
4-thiophen-2-ylbenzenesulfonyl chloride
C1=CSC(=C1)C2=CC=C(C=C2)S(=O)(=O)Cl
Packaging Glass Bottle
Melting Point 114°C to 115°C
Quantity 1g
Assay Percent Range 94% min. (HPLC)
Formula Weight 258.75
Percent Purity 95%
Chemical Name or Material 4-(2-Thienyl)benzenesulfonyl chloride

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Contact with water liberates toxic gas.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Danger