Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Thienyl)butyric Acid 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
4654-08-4
C11H14O3
194.23
MFCD00005463
YSSJQFONKASLKM-UHFFFAOYSA-N
5-4-hydroxyphenyl pentanoic acid,5-4-hydroxyphenyl valeric acid,pubchem16852,acmc-20am8d,5-4-hydroxyphenyl pentanoicacid,benzenepentanoic acid, 4-hydroxy
7016423
5-(4-hydroxyphenyl)pentanoic acid
C1=CC(=CC=C1CCCCC(=O)O)O
4654-08-4
C11H14O3
194.23
MFCD00005463
YSSJQFONKASLKM-UHFFFAOYSA-N
5-4-hydroxyphenyl pentanoic acid,5-4-hydroxyphenyl valeric acid,pubchem16852,acmc-20am8d,5-4-hydroxyphenyl pentanoicacid,benzenepentanoic acid, 4-hydroxy
7016423
5-(4-hydroxyphenyl)pentanoic acid
C1=CC(=CC=C1CCCCC(=O)O)O
Infrared Spectrum Authentic
Refractive Index 1.5310 to 1.5330 (20°C, 589nm)
Color Brown
Quantity 1g
Density 1.16g/mL
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 170.23
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 4-(2-Thienyl)butyric acid, 98%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger