Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(3-Bromo-4-fluorobenzyl)morpholine, 96%

Click to view available options
Quantity:
250mg
1g
5g
281652-25-3
C11H13BrFNO
274.133
MFCD01307672
KRXZMNYOZJCBKI-UHFFFAOYSA-N
4-3-bromo-4-fluorobenzyl morpholine,4-3-bromo-4-fluorophenyl methyl morpholine,cambridge id 5424905,2-fluoro-5-morpholin-4-ylmethyl-bromobenzene
782859
4-[(3-bromo-4-fluorophenyl)methyl]morpholine
C1COCCN1CC2=CC(=C(C=C2)F)Br
281652-25-3
C11H13BrFNO
274.133
MFCD01307672
KRXZMNYOZJCBKI-UHFFFAOYSA-N
4-3-bromo-4-fluorobenzyl morpholine,4-3-bromo-4-fluorophenyl methyl morpholine,cambridge id 5424905,2-fluoro-5-morpholin-4-ylmethyl-bromobenzene
782859
4-[(3-bromo-4-fluorophenyl)methyl]morpholine
C1COCCN1CC2=CC(=C(C=C2)F)Br
Quantity 1g
Formula Weight 274.13
Physical Form Solid
Percent Purity 96%
Chemical Name or Material 4-(3-Bromo-4-fluorobenzyl)morpholine

TSCA : No

Recommended Storage : Ambient temperatures