Alfa Aesar™ 4-(3-Piperidinyloxy)benzonitrile, 97%

Manufacturer: Alfa Aesar™ H5099503

Quantity 1g

 View more versions of this product

Catalog No. AAH5099503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 902837-34-7
    Molecular Formula C12H14N2O
    Formula Weight 202.26
    MDL Number MFCD08061105
    Chemical Name or Material 4-(3-Piperidinyloxy)benzonitrile

    Specifications

    Formula Weight 202.26
    MDL Number MFCD08061105
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Quantity 1g
    Molecular Formula C12H14N2O
    CAS 902837-34-7
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material 4-(3-Piperidinyloxy)benzonitrile
    Assay 97%

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit