Alfa Aesar™ 4(3H)-Pyrimidinone, 98%

Manufacturer: Alfa Aesar™ A1085906

Quantity 5g

 View more versions of this product

Catalog No. AAA1085906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 51953-17-4
    Molecular Formula C4H4N2O
    Formula Weight 96.09
    MDL Number MFCD00167178
    Beilstein 106975
    Synonym 4-Hydroxypyrimidine; 4-Pyrimidinol
    Chemical Name or Material 4 (3H)-Pyrimidinone

    Specifications

    Formula Weight 96.09
    Synonym 4-Hydroxypyrimidine; 4-Pyrimidinol
    Quantity 5g
    TSCA TSCA
    Molecular Formula C4H4N2O
    MDL Number MFCD00167178
    CAS 51953-17-4
    Beilstein 106975
    RTECS Number UW7350000
    Melting Point 164° to 167°C
    Chemical Name or Material 4 (3H)-Pyrimidinone
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit