Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
181219-01-2
C11H16BNO2
205.06
MFCD01319051
NLTIETZTDSJANS-UHFFFAOYSA-N
4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine,4-pyridineboronic acid pinacol ester,pyridine-4-boronic acid pinacol ester,4-pyridylboronic acid pinacol ester,pyridine-4-boronic acid, pinacol ester,4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine,pyridine-4-boronic pinacolate,pyridine-4-boronic acid pinacol cyclic ester,2-4-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,pyridine-4-boronic pinacol ester
2734648
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
B1(OC(C(O1)(C)C)(C)C)C2=CC=NC=C2
181219-01-2
C11H16BNO2
205.06
MFCD01319051
NLTIETZTDSJANS-UHFFFAOYSA-N
4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine,4-pyridineboronic acid pinacol ester,pyridine-4-boronic acid pinacol ester,4-pyridylboronic acid pinacol ester,pyridine-4-boronic acid, pinacol ester,4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine,pyridine-4-boronic pinacolate,pyridine-4-boronic acid pinacol cyclic ester,2-4-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,pyridine-4-boronic pinacol ester
2734648
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
B1(OC(C(O1)(C)C)(C)C)C2=CC=NC=C2
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point 153.0°C to 155.0°C
Color White
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Solubility Information Solubility in water: insoluble
Formula Weight 205.06
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS Signal Word: Warning

Recommended Storage : Refrigerator +4°C