Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 97%, Thermo Scientific Chemicals

Catalog Number p-2690482
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
181219-01-2
C11H16BNO2
205.06
MFCD01319051
NLTIETZTDSJANS-UHFFFAOYSA-N
4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 4-pyridineboronic acid pinacol ester, pyridine-4-boronic acid pinacol ester, 4-pyridylboronic acid pinacol ester, pyridine-4-boronic acid, pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine-4-boronic pinacolate, pyridine-4-boronic acid pinacol cyclic ester, 2-4-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, pyridine-4-boronic pinacol ester
2734648
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
CC1(C)OB(OC1(C)C)C1=CC=NC=C1
181219-01-2
C11H16BNO2
205.06
MFCD01319051
NLTIETZTDSJANS-UHFFFAOYSA-N
4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 4-pyridineboronic acid pinacol ester, pyridine-4-boronic acid pinacol ester, 4-pyridylboronic acid pinacol ester, pyridine-4-boronic acid, pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine-4-boronic pinacolate, pyridine-4-boronic acid pinacol cyclic ester, 2-4-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, pyridine-4-boronic pinacol ester
2734648
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
CC1(C)OB(OC1(C)C)C1=CC=NC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 153.0°C to 155.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 205.06
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Recommended Storage Refrigerator +4°C

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit