Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4,4-Trifluoro-1-(2-naphthyl)-1,3-butanedione 98.0+%, TCI America™

Click to view available options
Quantity:
5g
10g
25g
893-33-4
C14H9F3O2
266.219
MFCD00054501
WVVLURYIQCXPIV-UHFFFAOYSA-N
4,4,4-trifluoro-1-naphthalen-2-yl butane-1,3-dione,4,4,4-trifluoro-1-2-naphthyl-1,3-butanedione,1-2-naphthoyl-3,3,3-trifluoroacetone,4,4,4-trifluoro-1-2-naphthyl butane-1,3-dione,3-2-naphthoyl-1,1,1-trifluoroacetone,1,3-butanedione, 4,4,4-trifluoro-1-2-naphthalenyl,2-naphthoyltrifluoroacetone,1,3-butanedione, 4,4,4-trifluoro-1-2-naphthyl,4,4,4-trifluoro-3-oxo-2'-butyronaphthone,acmc-1bl5t
70179
4,4,4-trifluoro-1-naphthalen-2-ylbutane-1,3-dione
C1=CC=C2C=C(C=CC2=C1)C(=O)CC(=O)C(F)(F)F
893-33-4
C14H9F3O2
266.219
MFCD00054501
WVVLURYIQCXPIV-UHFFFAOYSA-N
4,4,4-trifluoro-1-naphthalen-2-yl butane-1,3-dione,4,4,4-trifluoro-1-2-naphthyl-1,3-butanedione,1-2-naphthoyl-3,3,3-trifluoroacetone,4,4,4-trifluoro-1-2-naphthyl butane-1,3-dione,3-2-naphthoyl-1,1,1-trifluoroacetone,1,3-butanedione, 4,4,4-trifluoro-1-2-naphthalenyl,2-naphthoyltrifluoroacetone,1,3-butanedione, 4,4,4-trifluoro-1-2-naphthyl,4,4,4-trifluoro-3-oxo-2'-butyronaphthone,acmc-1bl5t
70179
4,4,4-trifluoro-1-naphthalen-2-ylbutane-1,3-dione
C1=CC=C2C=C(C=CC2=C1)C(=O)CC(=O)C(F)(F)F
Quantity 25g
Color Undesignated
Melting Point 74°C
Formula Weight 266.22
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material 4,4,4-Trifluoro-1-(2-naphthyl)-1,3-butanedione