Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4,4-Trifluorobutylamine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
819-46-5
C4H8F3N
127.11
LAXWLCVPJLBABV-UHFFFAOYSA-N
4,4,4-trifluorobutylamine,4,4,4-trifluorobutyl-1-amine,1-butanamine,4,4,4-trifluoro,cf3ch2ch2ch2nh2,1-butanamine, 4,4,4-trifluoro,4,4,4-trifluoro-butylamine,4,4,4-trifluorobutyl-amine,4,4,4-trifluoro-1-butanamine,1,1,1-trifluorobutane-4-amine,1-amino-4,4,4-trifluorobutane
136645
4,4,4-trifluorobutan-1-amine
C(CC(F)(F)F)CN
819-46-5
C4H8F3N
127.11
LAXWLCVPJLBABV-UHFFFAOYSA-N
4,4,4-trifluorobutylamine,4,4,4-trifluorobutyl-1-amine,1-butanamine,4,4,4-trifluoro,cf3ch2ch2ch2nh2,1-butanamine, 4,4,4-trifluoro,4,4,4-trifluoro-butylamine,4,4,4-trifluorobutyl-amine,4,4,4-trifluoro-1-butanamine,1,1,1-trifluorobutane-4-amine,1-amino-4,4,4-trifluorobutane
136645
4,4,4-trifluorobutan-1-amine
C(CC(F)(F)F)CN
Boiling Point 90°C to 94°C
Packaging Glass bottle
Quantity 5g
Color Colorless
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 127.11
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 4, 4, 4-Trifluorobutylamine
Show More Show Less

GHS H Statement
Flammable liquid and vapour.
Causes severe skin burns and eye damage.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or ha

GHS Signal Word: Danger