Alfa Aesar™ 4,4',4''-Trimethoxytrityl alcohol, 95%

Manufacturer: Alfa Aesar™ B2451603

Quantity 1g

 View more versions of this product

Catalog No. AAB2451603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3010-81-9
    Molecular Formula C22H22O4
    Formula Weight 350.41
    MDL Number MFCD00014898
    Beilstein 1890559
    Synonym Tris(4-methoxyphenyl) carbinol; Tris(4-methoxyphenyl)methanol
    Chemical Name or Material 4,4',4''-Trimethoxytrityl alcohol

    Specifications

    Formula Weight 350.41
    Molecular Formula C22H22O4
    MDL Number MFCD00014898
    CAS 3010-81-9
    Beilstein 1890559
    Synonym Tris(4-methoxyphenyl) carbinol; Tris(4-methoxyphenyl)methanol
    Melting Point 76° to 82°C
    Quantity 1g
    Chemical Name or Material 4,4',4''-Trimethoxytrityl alcohol
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit