Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4,6-Dimethoxy[1.3.5]triazin-2-yl)-4-methylmorpholinium chloride, 97%, Acros Organics

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
3945-69-5
C10H17ClN4O3
276.72
MFCD03613550
BMTZEAOGFDXDAD-UHFFFAOYSA-M
dmtmm,4-4,6-dimethoxy-1,3,5-triazin-2-yl-4-methylmorpholinium chloride,4-4,6-dimethoxy-1,3,5-triazin-2-yl-4-methyl morpholinium chloride,dmt-mm,4-4,6-dimethoxy-1,3,5-triazine-2-yl-4-methyl morpholinium chloride,4-4,6-dimethoxy-1,3,5-triazin-2-yl-4-methylmorpholin-4-ium chloride,kunishima coupling reagent,mmtm,4-4,6-dimethoxy-1,3,5 triazin-2-yl-4-methyl-morpholin-4-ium chloride
2734059
4-(4,6-dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholin-4-ium;chloride
C[N+]1(CCOCC1)C2=NC(=NC(=N2)OC)OC.[Cl-]
3945-69-5
C10H17ClN4O3
276.72
MFCD03613550
BMTZEAOGFDXDAD-UHFFFAOYSA-M
dmtmm,4-4,6-dimethoxy-1,3,5-triazin-2-yl-4-methylmorpholinium chloride,4-4,6-dimethoxy-1,3,5-triazin-2-yl-4-methyl morpholinium chloride,dmt-mm,4-4,6-dimethoxy-1,3,5-triazine-2-yl-4-methyl morpholinium chloride,4-4,6-dimethoxy-1,3,5-triazin-2-yl-4-methylmorpholin-4-ium chloride,kunishima coupling reagent,mmtm,4-4,6-dimethoxy-1,3,5 triazin-2-yl-4-methyl-morpholin-4-ium chloride
2734059
4-(4,6-dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholin-4-ium;chloride
C[N+]1(CCOCC1)C2=NC(=NC(=N2)OC)OC.[Cl-]
Coupling reagent
Melting Point 118°C to 120°C
Quantity 5g
Packaging Glass bottle
Color White
Infrared Spectrum Authentic
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 276.72
Physical Form Powder or Chunks
Percent Purity 97%
Chemical Name or Material 4-(4, 6-Dimethoxy[1.3.5]triazin-2-yl)-4-methylmorpholinium chloride
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a POISON CENTER or doctor/

GHS Signal Word: Danger