Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4,6-Trimethyl-2-vinyl-1,3,2-dioxaborinane, 98%, stabilized, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass Bottle in Plastic Container
Glass bottle
4627-10-5
C8H15BO2
154.02
MFCD00066915
LFOAVJXWGLVDDI-UHFFFAOYNA-N
4,4,6-trimethyl-2-vinyl-1,3,2-dioxaborinane,vinylboronic acid 2-methyl-2,4-pentanediol ester,1,3,2-dioxaborinane,2-ethenyl-4,4,6-trimethyl,4,4,6-trimethyl-2-vinyl-1,3,2-dioxaborinane stabilized with thf,2-vinyl-4,4,6-trimethyl-2-bora-1,3-dioxane,vinylboronsäure-2-methyl-2,4-pentandiolester,1,3,2-dioxaborinane, 2-ethenyl-4,4,6-trimethyl,vinylboronsaure-2-methyl-2,4-pentandiolester,vinylboronic acid 2-methyl-2,4-pentanediol ester, contains phenothiazine as stabilizer
107310
2-ethenyl-4,4,6-trimethyl-1,3,2-dioxaborinane
B1(OC(CC(O1)(C)C)C)C=C
4627-10-5
C8H15BO2
154.02
MFCD00066915
LFOAVJXWGLVDDI-UHFFFAOYNA-N
4,4,6-trimethyl-2-vinyl-1,3,2-dioxaborinane,vinylboronic acid 2-methyl-2,4-pentanediol ester,1,3,2-dioxaborinane,2-ethenyl-4,4,6-trimethyl,4,4,6-trimethyl-2-vinyl-1,3,2-dioxaborinane stabilized with thf,2-vinyl-4,4,6-trimethyl-2-bora-1,3-dioxane,vinylboronsäure-2-methyl-2,4-pentandiolester,1,3,2-dioxaborinane, 2-ethenyl-4,4,6-trimethyl,vinylboronsaure-2-methyl-2,4-pentandiolester,vinylboronic acid 2-methyl-2,4-pentanediol ester, contains phenothiazine as stabilizer
107310
2-ethenyl-4,4,6-trimethyl-1,3,2-dioxaborinane
B1(OC(CC(O1)(C)C)C)C=C
Density 0.8800g/mL
Infrared Spectrum Authentic
Packaging Glass Bottle in Plastic Container
Quantity 250mg
Stabilizer 10% max. tetrahydrofuran
Color Colorless
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 154.02
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 4,4,6-Trimethyl-2-vinyl-1,3,2-dioxaborinane
Show More Show Less

GHS P Statement Flammable liquid and vapor. Suspected of causing cancer.

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. Obtain special instructions before use. Do not handle until all safety precautions have been read and understood.