Alfa Aesar™ 4-(4-Acetamidophenyl)-2-aminothiazole, 97%

Manufacturer: Alfa Aesar™ H2785406

Quantity 5g

 View more versions of this product

Catalog No. AAH2785406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 21674-96-4
    Molecular Formula C11H11N3OS
    Formula Weight 233.29
    MDL Number MFCD00207103
    Synonym 4-(4-Acetamidophenyl)-2-thiazolamine
    Chemical Name or Material 4-(4-Acetamidophenyl)-2-aminothiazole

    Specifications

    Formula Weight 233.29
    MDL Number MFCD00207103
    Synonym 4-(4-Acetamidophenyl)-2-thiazolamine
    Odor Odorless
    Molecular Formula C11H11N3OS
    CAS 21674-96-4
    Melting Point 248° to 252°C
    Quantity 5g
    Chemical Name or Material 4-(4-Acetamidophenyl)-2-aminothiazole
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit