Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4«-Azobis(4-cyanovaleric Acid) 97%, Acros Organics

Click to view available options
Quantity:
50g
2638-94-0
C12H16N4O4
280.284
VFXXTYGQYWRHJP-UHFFFAOYSA-N
4,4'-azobis 4-cyanovaleric acid,azobis cyanovaleric acid,4,4'-diazene-1,2-diyl bis 4-cyanopentanoic acid,4,4'-azobis 4-cyanopentanoic acid,acva,4,4-azobis 4-cyanovaleric acid,valeric acid, 4,4'-azobis 4-cyano,pentanoic acid, 4,4'-azobis 4-cyano,pentanoic acid, 4,4'-azobis 4-cyano,4,4'-azobis 4-cyano valeric acid acva
92938
4-[(4-carboxy-2-cyanobutan-2-yl)diazenyl]-4-cyanopentanoic acid
CC(CCC(=O)O)(C#N)N=NC(C)(CCC(=O)O)C#N
2638-94-0
C12H16N4O4
280.284
VFXXTYGQYWRHJP-UHFFFAOYSA-N
4,4'-azobis 4-cyanovaleric acid,azobis cyanovaleric acid,4,4'-diazene-1,2-diyl bis 4-cyanopentanoic acid,4,4'-azobis 4-cyanopentanoic acid,acva,4,4-azobis 4-cyanovaleric acid,valeric acid, 4,4'-azobis 4-cyano,pentanoic acid, 4,4'-azobis 4-cyano,pentanoic acid, 4,4'-azobis 4-cyano,4,4'-azobis 4-cyano valeric acid acva
92938
4-[(4-carboxy-2-cyanobutan-2-yl)diazenyl]-4-cyanopentanoic acid
CC(CCC(=O)O)(C#N)N=NC(C)(CCC(=O)O)C#N
Quantity 50g
CAS Min % 96.0
CAS Max % 100.0
Linear Formula [=NC(CH3)(CN)CH2CH2CO2H]2
Beilstein 04, IV, 3382
Formula Weight 280.27
Chemical Name or Material 4, 4'-Azobis(4-cyanovaleric acid), 97%

LIGHT SENSITIVE

EINECSNumber : 220-135-0

RTECSNumber : YV6190500

TSCA : TSCA